Display Info:
cookie.CK_ISO_CODE.value:us
cookie.CK_LANG_CODE.value:en
cookie.CK_DISPLAY_TYPE.value:
pageContext.request:org.apache.struts2.dispatcher.StrutsRequestWrapper@4d05a231
Request attributes: pageContext.request.scheme:http(not used)
syndicationProtocol:https
syndicationExtensionPrefix:
syndicationDisplayType:
Popular
Applications & Techniques
Shop All Products
Services
Support
Contact Us
Sign In
Quick Order
Search
Thermo Fisher Scientific
Search All
Search
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
Attention: Existing favorites lists will be migrated to Shared Lists by the end of 2020.
[Learn more about Shared Lists]
LOADING
See other PARD3 SIRNA Assays ›
Gene Symbol:
PARD3
Gene Name:
par-3 family cell polarity regulator
Gene Aliases:
ASIP, Baz, PAR3, PAR3alpha, PARD-3, PARD3A, PPP1R118, SE2-5L16, SE2-5LT1, SE2-5T2
Chromosome Location:
Chr.10: 34109560 - 34815325 on Build GRCh38
UniGene ID:
Hs.131489
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
28145
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PARD3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001184785.1
NP_001171714.1
20
3282
NM_001184786.1
NP_001171715.1
19
3243
NM_001184787.1
NP_001171716.1
20
3291
NM_001184788.1
NP_001171717.1
19
3153
NM_001184789.1
NP_001171718.1
19
3153
NM_001184790.1
NP_001171719.1
18
3021
NM_001184791.1
NP_001171720.1
17
3066
NM_001184792.1
NP_001171721.1
20
3282
NM_001184793.1
NP_001171722.1
20
3189
NM_001184794.1
NP_001171723.1
19
3153
NM_019619.3
NP_062565.2
20
3291
XM_005252528.4
XP_005252585.1
20
3270
XM_005252530.4
XP_005252587.1
19
3228
XM_005252531.4
XP_005252588.1
20
3183
XM_005252532.4
XP_005252589.1
19
2927
XM_005252534.4
XP_005252591.1
20
3270
XM_005252535.4
XP_005252592.1
19
3231
XM_011519569.2
XP_011517871.1
20
3279
XM_011519570.2
XP_011517872.1
19
3243
XM_011519571.2
XP_011517873.1
19
3237
XM_011519572.2
XP_011517874.1
19
3234
XM_011519573.2
XP_011517875.1
20
3192
XM_011519575.2
XP_011517877.1
19
2966
XM_011519576.2
XP_011517878.1
19
2937
XM_011519577.2
XP_011517879.1
18
3114
XM_011519578.2
XP_011517880.1
18
3112
XM_011519579.2
XP_011517881.1
18
2835
XM_011519581.2
XP_011517883.1
20
3282
XM_011519583.2
XP_011517885.1
20
3282
XM_017016430.1
XP_016871919.1
19
3225
GenBank mRNA
AB073671.1
-
17
2736
AF177228.1
-
13
1937
AF196185.1
-
20
3239
AF196186.1
-
20
3239
AF252293.1
-
18
2691
AF332592.1
-
19
2823
AF332593.1
-
19
2823
AF454057.1
-
18
2659
AF454058.1
-
18
3131
AF454059.1
-
15
2243
AF467002.1
-
20
2961
AF467003.1
-
20
2952
AF467004.1
-
20
2961
AF467005.1
-
17
2736
AF467006.1
-
19
2823
AK024668.1
-
7
934
AK025892.1
-
11
1729
AK027735.1
-
18
2486
BC011711.2
-
3
384
BC071566.1
-
19
3190
30344732
BM796605.1
-
3
385
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
Display Info:
cookie.CK_ISO_CODE.value:us
cookie.CK_LANG_CODE.value:en
cookie.CK_DISPLAY_TYPE.value:
pageContext.request.scheme:http(not used)
syndicationProtocol:https
syndicationExtensionPrefix:
syndicationDisplayType:
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-srch-2-prod-green:8080/10.253.226.220:8080.
git-commit: 0f6c802f6da5e59cd1b986a4dc444b4bcf3ac46d
git-url: http://victoria.invitrogen.com:8333/magellan/core.git
git-branch: origin/release/1.15.0-2021.01.02-1.0