Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Irf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033691.1 | NP_001028863.1 | 2 | 540 | ||
XM_006230540.1 | XP_006230602.1 | 3 | 639 | |||
XM_006230541.1 | XP_006230603.1 | 3 | 639 | |||
GenBank mRNA | BC104686.1 | - | 2 | 540 | 7442863 |