Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TMEM167B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322248.1 | NP_001309177.1 | 3 | 2141 | ||
NM_020141.3 | NP_064526.1 | 3 | 2040 | |||
NR_136242.1 | 4 | 2169 | ||||
NR_136243.1 | 2 | 1948 | ||||
GenBank mRNA | AK075191.1 | - | 3 | 2062 | ||
BC008739.2 | - | 3 | 2040 | 3346340 | ||
BC065186.1 | - | 3 | 2040 | 4642864 | ||
BC066918.1 | - | 3 | 2069 | 4826492 | ||
BC067116.1 | - | 3 | 2026 | 5532892 | ||
BC171849.1 | - | 3 | 3018 | 9054503 |