Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034151.1 | NP_001029323.1 | 6 | 970 | ||
GenBank mRNA | BC101399.1 | - | 6 | 970 | 7933358 |