Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BMP10 SIRNA Assays ›
Gene Symbol:
Bmp10
Gene Name:
bone morphogenetic protein 10
Gene Aliases:
-
Chromosome Location:
Chr.4: 119224906 - 119230534 on Build Rnor_6.0
UniGene ID:
Rn.118010
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
282345
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Bmp10
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001031824.1
NP_001026994.1
1
178
GenBank mRNA
AB158417.1
-
1
178
AB158418.1
-
1
178
Back To Top
More Information
Panther Classification:
Molecular Function -
growth factor
Gene Ontology Categories:
Function(s)
Process(es)
cell adhesion
heart development
adult heart development
negative regulation of endothelial cell migration
positive regulation of cardiac muscle hypertrophy
negative regulation of cardiac muscle hypertrophy
positive regulation of gene expression
positive regulation of pathway-restricted SMAD protein phosphorylation
negative regulation of cell growth
negative regulation of cell migration
BMP signaling pathway
activin receptor signaling pathway
regulation of apoptotic process
regulation of MAPK cascade
sarcomere organization
positive regulation of transcription, DNA-templated
atrial cardiac muscle tissue morphogenesis
ventricular cardiac muscle tissue morphogenesis
ventricular cardiac muscle cell development
regulation of cardiac muscle contraction
cardiac muscle cell proliferation
positive regulation of cardiac muscle cell proliferation
positive regulation of sarcomere organization
heart trabecula formation
pathway-restricted SMAD protein phosphorylation
SMAD protein signal transduction
regulation of cardiac muscle hypertrophy in response to stress
positive regulation of cell proliferation involved in heart morphogenesis
molecular_function
cytokine activity
transforming growth factor beta receptor binding
hormone activity
growth factor activity
telethonin binding
receptor serine/threonine kinase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-zxk44:80/100.66.78.82:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0