Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppapdc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034854.1 | NP_001030026.1 | 1 | 2119 | ||
GenBank mRNA | BC081970.1 | - | 1 | 2119 | 7125531 |