Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yipf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207633.2 | NP_997516.1 | 7 | 1330 | ||
GenBank mRNA | AK030473.1 | - | 7 | 1332 | ||
AK045453.1 | - | 7 | 1042 | |||
AK053562.1 | - | 7 | 1330 | |||
AK143152.1 | - | 7 | 900 | |||
BC048712.1 | - | 7 | 965 | 6328657 |