Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scgb2b20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009952.2 | NP_001009952.2 | 2, 3 | 259 | ||
GenBank mRNA | AF272844.1 | - | 2, 3 | 263 | ||
CK616187.1 | - | 2, 3 | 260 |
Gene Symbol: Scgb2b12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281508.1 | NP_001268437.1 | 2, 3 | 256 | ||
GenBank mRNA | CK616067.1 | - | 2, 3 | 264 |
Gene Symbol: Scgb2b19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199336.1 | NP_001186265.1 | 2, 3 | 262 |
Gene Symbol: Scgb2b7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198871.1 | NP_001185800.1 | 2, 3 | 256 | ||
GenBank mRNA | CK615801.1 | - | 2, 3 | 257 | ||
CK616903.1 | - | 2, 3 | 254 |