Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdzd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033222.3 | NP_001028394.2 | 5 | 1753 | ||
GenBank mRNA | AK041847.1 | - | 5 | 1207 | ||
AK145020.1 | - | 5 | 1753 | |||
BC141370.1 | - | 5 | 1658 | 9055983 | ||
BC141371.1 | - | 5 | 1658 | 9055984 |