Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2310003H01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027980.2 | NP_082256.2 | 4 | 1396 | ||
GenBank mRNA | AK082976.1 | - | 4 | 1397 | ||
AK147900.1 | - | 1485 | ||||
AK148234.1 | - | 4 | 1381 | |||
AK165631.1 | - | 3 | 1267 | |||
AK198382.1 | - | 2 | 298 | |||
BC034203.1 | - | 4 | 1267 | 4986832 |