Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rasgef1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029004.1 | NP_083280.1 | 2 | 307 | ||
GenBank mRNA | AK018593.1 | - | 2 | 296 | ||
AK039330.1 | - | 671 |