Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GPER1 SIRNA Assays ›
          Gene Symbol:
          GPER1
          Gene Name:
          G protein-coupled estrogen receptor 1
          Gene Aliases:
          CEPR, CMKRL2, DRY12, FEG-1, GPCR-Br, GPER, GPR30, LERGU, LERGU2, LyGPR, mER
          Chromosome Location:
          Chr.7: 1086807 - 1093815 on Build GRCh38
          UniGene ID:
          Hs.20961
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 284631

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: GPER1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001039966.1 NP_001035055.1 3 2945
          NM_001098201.1 NP_001091671.1 2 2618
          NM_001505.2 NP_001496.1 2 2821
          GenBank mRNA AF015257.1 - 2 2750
          BC011634.1 - 1 2152 4079700
          BC082766.1 - 2 1877 5419168
          U63917.1 - 2 2564

          Back To Top

          More Information


          Set Membership:

          G-protein coupled receptor

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of protein phosphorylation
          positive regulation of neurotransmitter secretion
          negative regulation of leukocyte activation
          inflammatory response
          cell cycle
          G-protein coupled receptor signaling pathway
          positive regulation of cytosolic calcium ion concentration
          positive regulation of cell proliferation
          negative regulation of cell proliferation
          positive regulation of adenylate cyclase activity involved in G-protein coupled receptor signaling pathway
          positive regulation of gene expression
          negative regulation of gene expression
          negative regulation of cell cycle process
          positive regulation of phosphatidylinositol 3-kinase signaling
          neuronal action potential
          apoptotic chromosome condensation
          nuclear fragmentation involved in apoptotic nuclear change
          positive regulation of cell migration
          intracellular steroid hormone receptor signaling pathway
          positive regulation of cAMP biosynthetic process
          mineralocorticoid receptor signaling pathway
          positive regulation of insulin secretion
          positive regulation of inositol trisphosphate biosynthetic process
          positive regulation of apoptotic process
          positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
          steroid hormone mediated signaling pathway
          positive regulation of MAPK cascade
          innate immune response
          negative regulation of fat cell differentiation
          positive regulation of epidermal growth factor receptor signaling pathway
          positive regulation of G-protein coupled receptor protein signaling pathway
          positive regulation of vasodilation
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of inflammatory response
          positive regulation of neurogenesis
          negative regulation of DNA metabolic process
          negative regulation of lipid biosynthetic process
          positive regulation of release of sequestered calcium ion into cytosol
          regulation of cytosolic calcium ion concentration
          negative regulation of protein kinase B signaling
          negative regulation of ERK1 and ERK2 cascade
          positive regulation of ERK1 and ERK2 cascade
          positive regulation of uterine smooth muscle contraction
          negative regulation of cell cycle arrest
          cellular response to glucose stimulus
          cellular response to tumor necrosis factor
          cellular response to peptide hormone stimulus
          cellular response to mineralocorticoid stimulus
          cellular response to estradiol stimulus
          positive regulation of establishment of protein localization to plasma membrane
          positive regulation of release of cytochrome c from mitochondria
          negative regulation of vascular smooth muscle cell proliferation
          positive regulation of endothelial cell apoptotic process
          positive regulation of cardiac vascular smooth muscle cell differentiation
          positive regulation of extrinsic apoptotic signaling pathway
          chromatin binding
          G-protein coupled receptor activity
          steroid binding
          protein binding
          drug binding
          mineralocorticoid receptor activity
          estrogen receptor activity
          steroid hormone binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-9nscr:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0