Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HLA-B SIRNA Assays ›
          Gene Symbol:
          HLA-B
          Gene Name:
          major histocompatibility complex, class I, B
          Gene Aliases:
          AS, B-4901, B-5001, Bw-47, Bw-50, HEL-S-83, HLA-B*45ZJ, HLA-B-3506, HLA-B-3905, HLA-B-5502, HLA-B-5602, HLA-B15, HLA-B39, HLA-B49, HLA-B50, HLA-B55, HLA-B59, HLA-B61, HLA-Cw, HLA-DRB1, HLAB, SPDA1
          Chromosome Location:
          Chr.6: 31353866 - 31357245 on Build GRCh38
          UniGene ID:
          Hs.654404
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 284640

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: HLA-B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_005514.7 NP_005505.2 4 870
          XM_011514557.1 XP_011512859.1 4 793
          GenBank mRNA AB008102.1 - 4 783
          AF006634.1 - 4 783
          AF016641.1 - 4 783
          AF022783.1 - 4 783
          AF026218.1 - 4 783
          AF130734.1 - 5 783
          AF170577.1 - 4 783
          AF189017.1 - 4 783
          AF389378.1 - 4 787
          AJ131118.1 - 4 783
          AJ251593.1 - 4 782
          AJ506054.1 - 1 164
          AJ535113.1 - 4 787
          AJ634528.1 - 4 783
          AK026595.1 - 5 806
          AK124160.1 - 4 1053
          AK129796.1 - 4 804
          AK130137.1 - 4 804
          AK130155.1 - 4 806
          AK130166.1 - 4 804
          AK130169.1 - 4 810
          AK130654.1 - 5 743
          AK290570.1 - 4 804
          AK292226.1 - 4 804
          AK300685.1 - 5 713
          AK313911.1 - 4 804
          AM711637.1 - 4 783
          AY007140.1 - 4 803
          AY221104.1 - 3 687
          AY569160.1 - 3 710
          BC007243.1 - 5 777
          BC013187.1 - 4 792 4344323
          BC091497.1 - 4 787 30386550
          BX647315.1 - 5 805
          D14343.1 - 4 747
          D25275.1 - 4 783
          D31816.1 - 4 783
          D49824.1 - 4 783
          D50290.1 - 4 783
          D50291.1 - 4 783
          D50292.1 - 4 783
          D50293.1 - 4 783
          D50294.1 - 4 783
          D50295.1 - 4 783
          D50296.1 - 4 783
          D50299.1 - 4 783
          D50300.1 - 4 783
          D83043.1 - 4 783
          D85761.1 - 5 783
          D85762.1 - 4 783
          D87665.1 - 5 774
          DQ336696.1 - 4 771
          DQ336697.1 - 4 777
          DQ336698.1 - 4 771
          EU048339.1 - 1 164
          EU668336.1 - 4 783
          FM205711.1 - 4 783
          HE663398.1 - 5 732
          HG327115.1 - 4 783
          HG327117.1 - 4 783
          HG327119.1 - 4 783
          HG327120.1 - 4 783
          HG514457.1 - 4 783
          KU204955.1 - 2 440
          L04695.1 - 4 783
          L04696.1 - 4 783
          L07743.1 - 4 783
          L07950.1 - 4 783
          L09735.1 - 4 783
          L09736.1 - 4 783
          L11570.1 - 4 783
          L11571.1 - 4 783
          L11603.1 - 4 783
          L11604.1 - 4 783
          L11666.1 - 4 783
          L15005.1 - 4 783
          L19923.1 - 4 783
          L19937.1 - 4 783
          L20089.1 - 4 778
          L20090.1 - 5 778
          L22027.1 - 4 783
          L32862.1 - 4 783
          L33922.1 - 4 783
          L37880.1 - 4 783
          L37881.1 - 4 783
          L38504.1 - 4 783
          L41214.1 - 5 783
          L41925.1 - 5 783
          L42345.1 - 4 783
          M12678.1 - 4 774
          M15470.1 - 4 774
          M16102.1 - 4 790
          M24033.1 - 4 783
          M27540.1 - 3 597
          M28203.1 - 4 772
          M28204.1 - 4 747
          M28205.1 - 5 783
          M32317.1 - 4 783
          M32318.1 - 5 783
          M32319.1 - 5 783
          M32320.1 - 4 783
          M62852.1 - 3 711
          M75138.1 - 4 783
          M77774.1 - 5 783
          M77775.1 - 4 783
          M77776.1 - 4 783
          M77777.1 - 4 783
          M77778.1 - 4 783
          M81798.1 - 4 783
          M83191.1 - 3 711
          M83192.1 - 3 711
          M83193.1 - 3 711
          M83194.1 - 3 711
          M83195.1 - 3 711
          M84380.1 - 4 783
          M84381.1 - 4 783
          M84382.1 - 4 783
          M84383.1 - 4 783
          M84384.1 - 4 783
          M84385.1 - 4 783
          M84694.1 - 4 783
          M86403.1 - 4 759
          U01848.1 - 4 783
          U03027.1 - 4 783
          U03698.1 - 4 783
          U03859.1 - 4 783
          U04244.1 - 4 783
          U04245.1 - 4 783
          U06697.1 - 3 710
          U06862.1 - 4 783
          U09864.1 - 4 783
          U09912.1 - 4 783
          U11261.1 - 4 783
          U11262.1 - 4 783
          U11263.1 - 4 783
          U11264.1 - 4 783
          U11265.1 - 4 783
          U11266.1 - 4 783
          U11267.1 - 4 783
          U11268.1 - 4 783
          U11269.1 - 4 783
          U14943.1 - 4 783
          U17107.1 - 4 783
          U17572.1 - 4 783
          U21052.1 - 4 783
          U21053.1 - 4 783
          U29057.1 - 4 783
          U30904.1 - 4 783
          U30936.1 - 4 783
          U31971.1 - 4 783
          U35431.1 - 4 783
          U35734.1 - 4 783
          U36392.1 - 4 783
          U36492.1 - 4 783
          U38800.1 - 4 783
          U50710.1 - 4 783
          U57966.1 - 4 783
          U63653.1 - 4 783
          U64801.1 - 4 783
          U70528.1 - 4 783
          U70529.1 - 4 783
          U75533.1 - 4 783
          U80945.1 - 4 783
          U88407.1 - 4 783
          V00527.1 - 1 46
          X61706.1 - 4 783
          X61708.1 - 5 783
          X61709.1 - 4 783
          X61710.1 - 4 783
          X64366.1 - 4 783
          X64454.1 - 4 783
          X73578.1 - 4 783
          X75953.1 - 5 783
          X81363.1 - 4 783
          X84725.1 - 4 783
          X90390.1 - 4 783
          X90391.1 - 4 783
          X91749.1 - 4 783
          Y08994.1 - 5 783
          Y08995.1 - 4 783
          Y09118.1 - 4 783
          Y13567.1 - 4 783
          Z15143.1 - 5 783
          Z22651.1 - 4 783
          Z33453.1 - 4 784

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          major histocompatibility complex protein

          Gene Ontology Categories:

          Function(s) Process(es)

          antigen processing and presentation of peptide antigen via MHC class I
          antigen processing and presentation of exogenous peptide antigen via MHC class I, TAP-dependent
          antigen processing and presentation of exogenous peptide antigen via MHC class I, TAP-independent
          viral process
          antigen processing and presentation
          regulation of immune response
          interferon-gamma-mediated signaling pathway
          type I interferon signaling pathway
          antigen binding
          peptide antigen binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0