Display Info:
cookie.CK_ISO_CODE.value:us
cookie.CK_LANG_CODE.value:en
cookie.CK_DISPLAY_TYPE.value:
pageContext.request:org.apache.struts2.dispatcher.StrutsRequestWrapper@59892d02
Request attributes: pageContext.request.scheme:http(not used)
syndicationProtocol:https
syndicationExtensionPrefix:
syndicationDisplayType:
Popular
Applications & Techniques
Shop All Products
Services
Support
Sign in
Quick Order
Search
Thermo Fisher Scientific
Search All
Search
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
Attention: Existing favorites lists will be migrated to Shared Lists by the end of 2020.
[Learn more about Shared Lists]
LOADING
See other SOX18 SIRNA Assays ›
Gene Symbol:
SOX18
Gene Name:
SRY-box 18
Gene Aliases:
HLTRS, HLTS
Chromosome Location:
Chr.20: 64047726 - 64049626 on Build GRCh38
UniGene ID:
Hs.8619
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
284782
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
siRNA Details
Display Info:
cookie.CK_ISO_CODE.value:us
cookie.CK_LANG_CODE.value:en
cookie.CK_DISPLAY_TYPE.value:
pageContext.request:org.apache.struts2.dispatcher.StrutsRequestWrapper@59892d02
Request attributes: pageContext.request.scheme:http(not used)
syndicationProtocol:https
syndicationExtensionPrefix:
syndicationDisplayType:
Popular
Applications & Techniques
Shop All Products
Services
Support
Sign in
Quick Order
Search
Thermo Fisher Scientific
Search All
Search
Unexpected Error
Please try again - we encountered an error while attempting to complete your request.
Contact
website technical assistance
if you see this message again.
Display Info:
cookie.CK_ISO_CODE.value:us
cookie.CK_LANG_CODE.value:en
cookie.CK_DISPLAY_TYPE.value:
pageContext.request.scheme:http(not used)
syndicationProtocol:https
syndicationExtensionPrefix:
syndicationDisplayType:
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-srch-3-prod-green:8080/10.253.229.154:8080.
git-commit: 0f6c802f6da5e59cd1b986a4dc444b4bcf3ac46d
git-url: http://victoria.invitrogen.com:8333/magellan/core.git
git-branch: origin/release/1.15.0-2021.01.02-1.0