Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FAM86B1 SIRNA Assays ›
          Gene Symbol:
          FAM86B1 FAM86B2
          Gene Name:
          family with sequence similarity 86 member B1
          family with sequence similarity 86 member B2
          Gene Aliases:
          -
          -
          Chromosome Location:
          Chr.8: 12182104 - 12194118 on Build GRCh38
          Chromosome Location:
          Chr.8: 12424419 - 12436418 on Build GRCh38
          UniGene ID:
          Hs.675799
          Hs.458413
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID 284851

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: FAM86B1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001083537.1 NP_001077006.1 4 407
          NR_003494.1 4 407
          XM_006716256.3 XP_006716319.1 5 502
          XM_006716257.3 XP_006716320.1 4 430
          XM_006716260.3 XP_006716323.1 3 316
          XM_006716262.1 XP_006716325.1 4 283
          XM_011543842.2 XP_011542144.1 4 357
          XM_011543843.2 XP_011542145.1 4 378
          XM_017013916.1 XP_016869405.1 4 406
          XM_017013917.1 XP_016869406.1 3 327
          XM_017013918.1 XP_016869407.1 3 299
          GenBank mRNA AK098138.1 - 4 374
          AK308513.1 - 3 391
          BC098427.1 - 4 457
          BE729618.1 - 4 349
          Gene Symbol: FAM86B2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001137610.1 NP_001131082.1 5 464
          XM_005273603.4 XP_005273660.1 4 437
          XM_005273604.4 XP_005273661.1 3 356
          XM_005273605.4 XP_005273662.1 3 257
          XM_005273607.2 XP_005273664.1 4 419
          XM_006716387.3 XP_006716450.1 4 458
          XM_011544618.2 XP_011542920.1 5 509
          XM_011544619.2 XP_011542921.1 4 476
          XM_011544622.2 XP_011542924.1 4 378
          XM_017013740.1 XP_016869229.1 4 357
          XM_017013741.1 XP_016869230.1 3 294
          XM_017013742.1 XP_016869231.1 3 276
          GenBank mRNA AK291001.1 - 4 419
          AK297741.1 - 3 305
          AK298940.1 - 2 281
          AK309984.1 - 5 517

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          methylation
          methyltransferase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-7kcm7:80/100.66.76.55:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0