Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DMKN SIRNA Assays ›
Gene Symbol:
DMKN
Gene Name:
dermokine
Gene Aliases:
UNQ729, ZD52F10
Chromosome Location:
Chr.19: 35497217 - 35513678 on Build GRCh38
UniGene ID:
Hs.417795
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
284866
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DMKN
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001035516.3
NP_001030593.1
6
519
NM_001126056.2
NP_001119528.2
18
1819
NM_001126059.2
NP_001119531.1
12
870
NM_001190347.1
NP_001177276.1
17
1771
NM_001308380.1
NP_001295309.1
13
933
NM_001308383.1
NP_001295312.1
11
840
NM_033317.4
NP_201574.3
16
1852
NR_033746.1
14
960
XM_006723477.1
XP_006723540.1
16
1049
XM_006723484.1
XP_006723547.1
14
959
XM_006723489.1
XP_006723552.1
16
1035
XM_006723493.2
XP_006723556.1
14
940
XM_006723494.2
XP_006723557.1
14
922
XM_006723503.2
XP_006723566.1
16
968
XM_011527494.2
XP_011525796.1
22
2058
XM_011527495.2
XP_011525797.1
23
2093
XM_011527496.2
XP_011525798.1
21
2022
XM_011527497.2
XP_011525799.1
21
2016
XM_011527498.2
XP_011525800.1
22
2051
XM_011527499.2
XP_011525801.1
21
2013
XM_011527500.2
XP_011525802.1
21
2004
XM_011527501.2
XP_011525803.1
21
1998
XM_011527502.2
XP_011525804.1
20
1968
XM_011527503.2
XP_011525805.1
20
1962
XM_011527504.2
XP_011525806.1
20
1944
XM_011527505.2
XP_011525807.1
19
1917
XM_011527506.2
XP_011525808.1
19
1908
XM_011527507.2
XP_011525809.1
18
1872
XM_011527508.2
XP_011525810.1
18
1857
XM_011527509.2
XP_011525811.1
17
1815
XM_011527510.2
XP_011525812.1
17
1812
XM_011527513.1
XP_011525815.1
13
908
XM_011527514.1
XP_011525816.1
12
848
XM_017027475.1
XP_016882964.1
14
953
XM_017027476.1
XP_016882965.1
12
876
XM_017027477.1
XP_016882966.1
12
760
GenBank mRNA
AF086315.1
-
6
510
AK096215.1
-
12
1499
AL832080.1
-
6
520
BC004493.2
-
14
842
BC011886.2
-
14
889
BC035311.1
-
16
1846
4752921
BM923917.1
-
13
855
EF158323.1
-
12
851
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
cornified envelope assembly
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-qfwvt:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0