Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PKM SIRNA Assays ›
          Gene Symbol:
          PKM
          Gene Name:
          pyruvate kinase, muscle
          Gene Aliases:
          CTHBP, HEL-S-30, OIP3, PK3, PKM2, TCB, THBP1
          Chromosome Location:
          Chr.15: 72199029 - 72231624 on Build GRCh38
          UniGene ID:
          Hs.534770
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 286

          Purification
          Size
          Availability Inventoried
          Catalog # AM51331
          Price 6,301.00
          Your Price
          Online offer:
          6301.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PKM
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001206796.2 NP_001193725.1 5 1213
          NM_001206798.2 NP_001193727.1 4 901
          NM_001206799.1 NP_001193728.1 4 543
          NM_001316318.1 NP_001303247.1 4 1096
          NM_002654.5 NP_002645.3 4 876
          NM_182470.3 NP_872270.1 4 1092
          NM_182471.3 NP_872271.1 4 876
          XM_005254443.1 XP_005254500.1 5 600
          XM_005254445.4 XP_005254502.1 4 437
          XM_006720570.1 XP_006720633.1 5 697
          XM_011521670.2 XP_011519972.1 4 662
          XM_017022313.1 XP_016877802.1 4 437
          GenBank mRNA AK092369.1 - 4 626
          AK222927.1 - 4 442
          AK294315.1 - 5 702
          AK300800.1 - 4 467
          AK307855.1 - 5 443
          AK308264.1 - 4 467
          AK312253.1 - 4 442
          BC000481.2 - 4 458 2964687
          BC007640.1 - 4 442 3533876
          BC007952.2 - 4 454 4299213
          BC012811.2 - 4 454 2958817
          BC023592.2 - 4 404
          BC035198.1 - 4 408 3859987
          BC096823.1 - 4 643
          EU794631.1 - 4 595
          M23725.1 - 4 463
          M26252.1 - 4 443

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          liver development
          ATP biosynthetic process
          response to nutrient
          response to gravity
          programmed cell death
          response to muscle inactivity
          organ regeneration
          response to insulin
          pyruvate biosynthetic process
          skeletal muscle tissue regeneration
          canonical glycolysis
          cell-cell adhesion
          magnesium ion binding
          pyruvate kinase activity
          protein binding
          ATP binding
          kinase activity
          MHC class II protein complex binding
          potassium ion binding
          ADP binding
          poly(A) RNA binding
          cadherin binding involved in cell-cell adhesion

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0