Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kng1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009628.1 | NP_001009628.1 | 7 | 871 | ||
GenBank mRNA | BC088161.1 | - | 7 | 871 | 7369906 | |
FQ211311.1 | - | 5 | 432 | |||
FQ219641.1 | - | 5 | 434 | |||
M11661.1 | - | 7 | 769 | |||
M11885.1 | - | 2 | 69 |