Search
Search

Abhd17b| Gene Symbol: Abhd17b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014028.1 | NP_001014050.1 | 2 | 700 | ||
| XM_006231162.2 | XP_006231224.1 | 2 | 790 | |||
| GenBank mRNA | BC079229.1 | - | 2 | 700 | 7132456 | |
| FQ227098.1 | - | 1 | 48 | |||