Search
Search

Ccdc17| Gene Symbol: Ccdc17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014067.2 | NP_001014089.2 | 12 | 1909 | ||
| XM_006238702.2 | XP_006238764.1 | 11 | 2139 | |||
| XM_006238703.2 | XP_006238765.1 | 12 | 2073 | |||
| XM_006238704.2 | XP_006238766.1 | 12 | 2067 | |||
| XM_006238705.2 | XP_006238767.1 | 11 | 1962 | |||
| XM_006238706.2 | XP_006238768.1 | 7 | 1683 | |||
| XM_006238707.2 | XP_006238769.1 | 13 | 2001 | |||
| GenBank mRNA | BC087059.1 | - | 10 | 2153 | ||