Search
Search

TUBGCP6| Gene Symbol: TUBGCP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020461.3 | NP_065194.2 | 12 | 2643 | ||
| GenBank mRNA | AB051456.1 | - | 12 | 2037 | ||
| AF272887.1 | - | 12 | 2236 | |||
| AK000636.1 | - | 4 | 442 | |||
| AL137665.1 | - | 5 | 460 | |||
| AL832617.2 | - | 12 | 2664 | |||
| BC113849.1 | - | 2 | 172 | |||
| BC114457.1 | - | 2 | 172 | |||
| BC127614.1 | - | 2 | 172 | |||
| BC144488.1 | - | 12 | 2235 | 9053015 | ||
| BC144489.1 | - | 12 | 2235 | |||
| BC150613.1 | - | 12 | 2235 | 9056983 | ||
| BC151209.1 | - | 12 | 2625 | 40147276 | ||