Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANP32B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006401.2 | NP_006392.1 | 2 | 462 | ||
GenBank mRNA | AK223454.1 | - | 2 | 329 | ||
AK313733.1 | - | 2 | 317 | |||
BC013003.2 | - | 2 | 294 | 3010189 | ||
BC019658.1 | - | 2 | 301 | 4478860 | ||
U70439.1 | - | 2 | 323 | |||
Y07569.1 | - | 2 | 168 | |||
Y07570.1 | - | 2 | 297 | |||
Y07969.1 | - | 2 | 337 |