Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspa14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015765.2 | NP_056580.2 | 3 | 317 | ||
GenBank mRNA | AK031489.1 | - | 256 | |||
AK031579.1 | - | 3 | 262 | |||
AK088996.1 | - | 3 | 247 | |||
AK145937.1 | - | 3 | 271 | |||
AK150000.1 | - | 3 | 293 | |||
AK153397.1 | - | 3 | 279 | |||
BC002056.1 | - | 3 | 245 | 3493119 | ||
BY147434.1 | - | 3 | 317 | |||
U08215.1 | - | 3 | 277 |