siRNA Details
Gene Symbol: Sox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009233.3 | NP_033259.2 | 1 | 856 | ||
GenBank mRNA | AB108672.1 | - | 1 | 53 |

More Information
Other Species Targeted

Panther Classification:
