Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Enterprise Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SIX3 SIRNA Assays ›
          Gene Symbol:
          Six3
          Gene Name:
          sine oculis-related homeobox 3
          Gene Aliases:
          E130112M24Rik, Six3a, Six3alpha, Six3b, Six3beta
          Chromosome Location:
          Chr.17: 85620834 - 85626191 on Build GRCm38
          UniGene ID:
          Mm.370208
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 287460

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Six3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_011381.4 NP_035511.2 1 642
          GenBank mRNA BC094426.1 - 445 6855602
          BC098096.1 - 1 262 6402835
          BF467709.1 - 1 280
          D83144.1 - 1 903
          D83145.1 - 1 228
          X90871.1 - 417

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Human: NM_005413, AL162671, AJ012611, BC030289.
          This siRNA also targets the following transcripts in Rat: NM_023990, AB030833.

          Panther Classification:

          Molecular Function -

          homeodomain transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          protein import into nucleus, translocation
          eye development
          epithelial cell maturation
          lens development in camera-type eye
          optic vesicle morphogenesis
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          transcription from RNA polymerase II promoter
          protein import into nucleus
          multicellular organism development
          brain development
          proximal/distal axis specification
          neuroblast differentiation
          diencephalon development
          telencephalon development
          forebrain anterior/posterior pattern specification
          forebrain dorsal/ventral pattern formation
          cell proliferation in forebrain
          telencephalon regionalization
          pituitary gland development
          negative regulation of Wnt signaling pathway
          regulation of cell proliferation
          camera-type eye development
          negative regulation of neuron differentiation
          negative regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          circadian behavior
          lens induction in camera-type eye
          regulation of neural retina development
          lens fiber cell differentiation
          neuroblast migration
          regulation of cell cycle phase transition
          regulation of neuroblast proliferation
          apoptotic process involved in development
          lens fiber cell apoptotic process
          regulation of neural precursor cell proliferation
          RNA polymerase II distal enhancer sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II distal enhancer sequence-specific binding
          transcription corepressor binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
          transcription coactivator activity
          receptor binding
          protein binding
          histone deacetylase binding
          sequence-specific DNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-2gfbh:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0