Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tuba1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011654.2 | NP_035784.1 | 4 | 1498 | ||
GenBank mRNA | AK075955.1 | - | 4 | 1545 | ||
AK137885.1 | - | 4 | 1542 | |||
AK150128.1 | - | 4 | 1545 | |||
AK150179.1 | - | 4 | 1545 | |||
AK150968.1 | - | 4 | 1544 | |||
AK151199.1 | - | 4 | 1542 | |||
AK151809.1 | - | 4 | 1542 | |||
AK152298.1 | - | 4 | 1542 | |||
AK153064.1 | - | 4 | 1542 | |||
AK153254.1 | - | 4 | 1542 | |||
AK158958.1 | - | 4 | 1674 | |||
AK164428.1 | - | 4 | 1545 | |||
AK168770.1 | - | 4 | 1542 | |||
AK169075.1 | - | 4 | 1542 | |||
AK169092.1 | - | 4 | 1545 | |||
AK169130.1 | - | 4 | 1545 | |||
AK169665.1 | - | 4 | 1545 | |||
AK191172.1 | - | 1 | 352 | |||
AK192129.1 | - | 1 | 331 | |||
AK192775.1 | - | 1 | 349 | |||
AK193004.1 | - | 1 | 310 | |||
AK194180.1 | - | 1 | 310 | |||
AK194222.1 | - | 1 | 304 | |||
AK194839.1 | - | 1 | 305 | |||
AK196308.1 | - | 1 | 319 | |||
AK197610.1 | - | 1 | 352 | |||
AK197770.1 | - | 1 | 353 | |||
AK199828.1 | - | 1 | 299 | |||
AK200029.1 | - | 1 | 323 | |||
AK200084.1 | - | 1 | 352 | |||
AK203041.1 | - | 1 | 275 | |||
AK207058.1 | - | 1 | 309 | |||
AK211470.1 | - | 1 | 320 | |||
BC002219.1 | - | 4 | 1532 | 3490568 | ||
BC008117.1 | - | 4 | 1525 | 3498312 | ||
BC020390.1 | - | 1385 | ||||
BC063777.1 | - | 4 | 1495 | 4458530 | ||
BC083120.1 | - | 4 | 1523 | 6311394 | ||
BC108337.1 | - | 4 | 1524 | 4167097 | ||
BC108394.1 | - | 4 | 1498 | 6505244 | ||
M13446.1 | - | 4 | 1522 | |||
M28727.1 | - | 2 | 1146 |