Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH2B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020979.4 | NP_066189.3 | 2 | 798 | ||
XM_005276975.4 | XP_005277032.1 | 2 | 1334 | |||
XM_005276976.3 | XP_005277033.1 | 2 | 1326 | |||
XM_005276977.4 | XP_005277034.1 | 2 | 833 | |||
XM_005276979.3 | XP_005277036.1 | 2 | 749 | |||
XM_011515740.2 | XP_011514042.1 | 2 | 1580 | |||
XM_017011680.1 | XP_016867169.1 | 2 | 834 | |||
XM_017011681.1 | XP_016867170.1 | 2 | 752 | |||
GenBank mRNA | AB000520.1 | - | 2 | 699 | ||
BI261466.1 | - | 2 | 619 |