Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H2-Ab1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207105.3 | NP_996988.2 | 2 | 257 | ||
GenBank mRNA | AK077436.1 | - | 2 | 245 | ||
AK155573.1 | - | 2 | 235 | |||
AK156899.1 | - | 2 | 235 | |||
AK170370.1 | - | 2 | 239 | |||
AK171005.1 | - | 2 | 235 | |||
AK171211.1 | - | 2 | 239 | |||
AK171281.1 | - | 2 | 235 | |||
AK171290.1 | - | 2 | 235 | |||
AK171298.1 | - | 2 | 235 | |||
AK172409.1 | - | 2 | 235 | |||
AY303784.1 | - | 1 | 90 | |||
AY303785.1 | - | 1 | 81 | |||
AY452202.1 | - | 2 | 172 | |||
BC008168.1 | - | 2 | 255 | 3257494 | ||
BC010322.1 | - | 2 | 223 | 2651058 | ||
BY205244.1 | - | 2 | 276 | |||
M13539.1 | - | 2 | 172 | |||
M15848.1 | - | 2 | 208 |