Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ITGB4 SIRNA Assays ›
          Gene Symbol:
          ITGB4
          Gene Name:
          integrin subunit beta 4
          Gene Aliases:
          CD104, GP150
          Chromosome Location:
          Chr.17: 75721328 - 75757819 on Build GRCh38
          UniGene ID:
          Hs.632226
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 288542

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ITGB4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000213.4 NP_000204.3 30, 31 4086
          NM_001005619.1 NP_001005619.1 29, 30 3799
          NM_001005731.2 NP_001005731.1 30, 31 4086
          NM_001321123.1 NP_001308052.1 30, 31 4037
          XM_005257309.2 XP_005257366.1 30, 31 3856
          XM_005257311.4 XP_005257368.1 30, 31 4075
          XM_006721866.3 XP_006721929.1 30, 31 4002
          XM_006721867.3 XP_006721930.1 30, 31 3999
          XM_006721868.3 XP_006721931.1 30, 31 3996
          XM_006721870.3 XP_006721933.1 30, 31 3992
          XM_011524751.2 XP_011523053.1 30, 31 4000
          XM_011524752.2 XP_011523054.1 15, 16 2285
          GenBank mRNA AB208913.1 - 30, 31 3888
          BC118916.1 - 30, 31 3848 40073279
          BC126411.1 - 30, 31 3907 8992127
          BC143738.1 - 30, 31 3907 9052252
          BC143742.1 - 30, 31 3858 9052256
          X51841.1 - 30, 31 3917
          X52186.1 - 29, 30 3800
          X53587.1 - 30, 31 3978

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          integrin

          Gene Ontology Categories:

          Function(s) Process(es)

          autophagy
          cell adhesion
          cell-matrix adhesion
          integrin-mediated signaling pathway
          response to wounding
          extracellular matrix organization
          hemidesmosome assembly
          nail development
          skin development
          mesodermal cell differentiation
          digestive tract development
          cell motility
          renal system development
          amelogenesis
          G-protein coupled receptor binding
          receptor activity
          protein binding
          insulin-like growth factor I binding
          neuregulin binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-pnd5g:80/100.66.79.83:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline