siRNA Details
Gene Symbol: POU3F3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006236.2 | NP_006227.1 | 1 | 1489 |

More Information
Other Species Targeted


Gene Symbol: POU3F3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006236.2 | NP_006227.1 | 1 | 1489 |