Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GPHN SIRNA Assays ›
Gene Symbol:
GPHN
Gene Name:
gephyrin
Gene Aliases:
GEPH, GPH, GPHRYN, HKPX1, MOCODC
Chromosome Location:
Chr.14: 66507407 - 67735831 on Build GRCh38
UniGene ID:
Hs.208765
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
28961
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: GPHN
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001024218.1
NP_001019389.1
6, 7
1585
NM_020806.4
NP_065857.1
6, 7
1585
XM_005267254.3
XP_005267311.1
6, 7
880
XM_011536340.2
XP_011534642.1
7, 8
1097
XM_011536342.2
XP_011534644.1
6, 7
1058
XM_011536343.2
XP_011534645.1
7, 8
1097
XM_011536344.2
XP_011534646.1
7, 8
1097
XM_011536345.2
XP_011534647.1
7, 8
1097
XM_011536346.2
XP_011534648.1
6, 7
859
XM_011536347.2
XP_011534649.1
7, 8
583
XM_017020913.1
XP_016876402.1
7, 8
1097
XM_017020914.1
XP_016876403.1
7, 8
1097
XM_017020915.1
XP_016876404.1
7, 8
1097
XM_017020916.1
XP_016876405.1
6, 7
1058
XM_017020917.1
XP_016876406.1
7, 8
1097
XM_017020918.1
XP_016876407.1
7, 8
1097
XM_017020919.1
XP_016876408.1
6, 7
1058
XM_017020920.1
XP_016876409.1
7, 8
1020
XM_017020921.1
XP_016876410.1
7, 8
1160
XM_017020922.1
XP_016876411.1
8, 9
626
XM_017020923.1
XP_016876412.1
7, 8
1160
XM_017020924.1
XP_016876413.1
7, 8
1097
XM_017020925.1
XP_016876414.1
7, 8
1097
XM_017020926.1
XP_016876415.1
7, 8
1097
GenBank mRNA
AB037806.1
-
6, 7
1585
AF272663.1
-
6, 7
1585
AJ272033.1
-
6, 7
464
AJ272343.1
-
6, 7
504
AK025169.1
-
6, 7
800
AK294532.1
-
7, 8
527
AK295577.1
-
4, 5
362
AK297447.1
-
5, 6
386
AK315569.1
-
6, 7
834
BC030016.2
-
6, 7
476
4826821
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
Mo-molybdopterin cofactor biosynthetic process
molybdenum incorporation into molybdenum-molybdopterin complex
molybdopterin cofactor biosynthetic process
glycine receptor clustering
gamma-aminobutyric acid receptor clustering
protein binding
ATP binding
metal ion binding
molybdopterin adenylyltransferase activity
molybdopterin molybdotransferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-9fjjm:80/10.239.165.64:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0