Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIRT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242827.1 | NP_001229756.1 | 6 | 738 | ||
NM_012241.4 | NP_036373.1 | 7 | 894 | |||
NM_031244.3 | NP_112534.1 | 7 | 872 | |||
XM_005248968.4 | XP_005249025.1 | 7 | 809 | |||
XM_011514444.2 | XP_011512746.1 | 8 | 943 | |||
XM_017010620.1 | XP_016866109.1 | 6 | 772 | |||
XM_017010621.1 | XP_016866110.1 | 7 | 809 | |||
XM_017010622.1 | XP_016866111.1 | 8 | 1002 | |||
XM_017010623.1 | XP_016866112.1 | 8 | 1132 | |||
XM_017010624.1 | XP_016866113.1 | 6 | 772 | |||
XM_017010625.1 | XP_016866114.1 | 6 | 772 | |||
GenBank mRNA | AF083110.1 | - | 7 | 855 | ||
AJ420496.1 | - | 7 | 801 | |||
AK000355.1 | - | 7 | 800 | |||
AK294162.1 | - | 6 | 666 | |||
BC000126.1 | - | 7 | 878 | 3352592 | ||
BX538030.1 | - | 6 | 685 |