Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Food and Beverage
Lab Solutions
Pharma and Biopharma
Real-Time PCR
Semiconductor Analysis
Clinical and Diagnostics
Digital Solutions
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
See all services
Help and Support
Order Help
Digital Solutions
Product Support
Technical Information
Training and Education
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RACGAP1 SIRNA Assays ›
Gene Symbol:
RACGAP1
Gene Name:
Rac GTPase activating protein 1
Gene Aliases:
CYK4, HsCYK-4, ID-GAP, MgcRacGAP
Chromosome Location:
Chr.12: 49989162 - 50033136 on Build GRCh38
UniGene ID:
Hs.505469
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
289673
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RACGAP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001126103.2
NP_001119575.1
17
2055
NM_001126104.2
NP_001119576.1
15
1935
NM_001319999.1
NP_001306928.1
15
1865
NM_001320000.1
NP_001306929.1
16
1939
NM_001320001.1
NP_001306930.1
15
1931
NM_001320002.1
NP_001306931.1
15
1890
NM_001320003.1
NP_001306932.1
15
1861
NM_001320004.1
NP_001306933.1
15
1812
NM_001320005.1
NP_001306934.1
14
1776
NM_001320006.1
NP_001306935.1
16
1935
NM_001320007.1
NP_001306936.1
14
1662
NM_013277.4
NP_037409.2
17
2044
XM_006719359.1
XP_006719422.1
15
1763
XM_011538238.1
XP_011536540.1
15
1720
XM_011538243.1
XP_011536545.1
15
1840
XM_017019220.1
XP_016874709.1
15
2037
XM_017019221.1
XP_016874710.1
14
1681
XM_017019222.1
XP_016874711.1
14
1636
XM_017019223.1
XP_016874712.1
14
1677
XM_017019224.1
XP_016874713.1
14
1607
XM_017019225.1
XP_016874714.1
14
1516
XM_017019226.1
XP_016874715.1
14
1576
XM_017019227.1
XP_016874716.1
14
1597
GenBank mRNA
AB030251.1
-
15
1771
AB040911.1
-
13
1516
AK000733.1
-
15
1778
AK225901.1
-
14
1706
AK315785.1
-
17
1992
AL136794.1
-
17
1925
BC024144.2
-
15
1757
BC032754.1
-
15
1762
5583315
CR533565.1
-
14
1701
Back To Top
More Information
Panther Classification:
Molecular Function -
GTPase-activating protein
Gene Ontology Categories:
Function(s)
Process(es)
mitotic cytokinesis
actomyosin contractile ring assembly
retrograde vesicle-mediated transport, Golgi to ER
microtubule-based movement
spermatogenesis
neuroblast proliferation
sulfate transport
embryo development
antigen processing and presentation of exogenous peptide antigen via MHC class II
positive regulation of cytokinesis
intracellular signal transduction
positive regulation of GTPase activity
regulation of small GTPase mediated signal transduction
mitotic spindle midzone assembly
regulation of attachment of spindle microtubules to kinetochore
GTPase activator activity
protein binding
phosphatidylinositol-3,4,5-trisphosphate binding
microtubule binding
protein kinase binding
alpha-tubulin binding
gamma-tubulin binding
metal ion binding
beta-tubulin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0