Search
Search

GNG13| Gene Symbol: GNG13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016541.2 | NP_057625.1 | 2 | 125 | ||
| GenBank mRNA | AB030207.1 | - | 2 | 104 | ||
| AK312139.1 | - | 2 | 126 | |||
| AY029486.1 | - | 2 | 91 | |||
| BC093760.1 | - | 1 | 56 | 7939605 | ||
| DA111448.1 | - | 2 | 126 | |||