Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HES2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019089.4 | NP_061962.2 | 4 | 684 | ||
GenBank mRNA | AK091122.1 | - | 4 | 684 | ||
BC132698.1 | - | 4 | 647 | 40146720 | ||
BC136963.1 | - | 4 | 647 | 9020955 | ||
BC142687.1 | - | 4 | 664 | 40148619 |