Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOC4L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024078.2 | NP_076983.1 | 15 | 1632 | ||
XM_017019960.1 | XP_016875449.1 | 14 | 1586 | |||
GenBank mRNA | BC001191.1 | - | 15 | 1611 | 3354945 | |
BC007893.2 | - | 15 | 1504 | |||
BC071958.1 | - | 15 | 1575 | 6597539 |