Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284355.2 | NP_001271284.1 | 6 | 2658 | ||
NM_133463.3 | NP_597720.1 | 7 | 2883 | |||
XM_011515150.2 | XP_011513452.1 | 7 | 2828 | |||
GenBank mRNA | AB075830.1 | - | 7 | 2876 | ||
BC146753.1 | - | 7 | 2876 | 9007189 |