Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PTGER3 SIRNA Assays ›
          Gene Symbol:
          PTGER3
          Gene Name:
          prostaglandin E receptor 3
          Gene Aliases:
          EP3, EP3-I, EP3-II, EP3-III, EP3-IV, EP3-VI, EP3e, PGE2-R
          Chromosome Location:
          Chr.1: 70852353 - 71047814 on Build GRCh38
          UniGene ID:
          Hs.445000
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 290305

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PTGER3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001126044.1 NP_001119516.1 2 1202
          NM_198714.1 NP_942007.1 2 1202
          NM_198715.2 NP_942008.1 2 1202
          NM_198716.1 NP_942009.1 2 1202
          NM_198717.1 NP_942010.1 2 1202
          NM_198718.1 NP_942011.1 2 1202
          NM_198719.1 NP_942012.1 2 1202
          NR_028292.1 2 1202
          NR_028293.1 2 1202
          NR_028294.1 2 1202
          XM_011541810.2 XP_011540112.1 2 1208
          GenBank mRNA AB451481.1 - 2 971
          AK290634.1 - 2 1068
          AK315825.1 - 2 1130
          AY387856.1 - 2 971
          AY429108.1 - 2 971
          BC024229.1 - 2 1212 4660371
          BC118578.1 - 2 992 40069637
          BC118659.1 - 2 992 40069636
          D38297.1 - 2 1202
          D38298.1 - 2 1202
          D38299.1 - 2 1202
          D38300.1 - 2 1202
          D38301.1 - 2 1202
          D86097.1 - 2 1202
          D86098.1 - 2 1202
          EF534325.1 - 2 126
          L26976.1 - 2 1110
          L27488.1 - 2 1188
          L27489.1 - 2 1188
          L27490.1 - 2 1166
          S69200.1 - 2 1188
          S69326.1 - 2 1167
          U13214.1 - 2 1027
          U13215.1 - 2 1027
          U13216.1 - 2 1027
          U13217.1 - 2 1027
          U13218.1 - 2 1027
          X83857.1 - 2 1178
          X83858.1 - 2 1178
          X83859.1 - 2 1178
          X83860.1 - 2 1178
          X83861.1 - 2 1178
          X83862.1 - 2 1178
          X83863.1 - 2 1178

          Back To Top

          More Information


          Set Membership:

          G-protein coupled receptor

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          inflammatory response
          G-protein coupled receptor signaling pathway
          adenylate cyclase-activating G-protein coupled receptor signaling pathway
          phospholipase C-activating G-protein coupled receptor signaling pathway
          positive regulation of cytosolic calcium ion concentration
          female pregnancy
          cell death
          positive regulation of gene expression
          negative regulation of norepinephrine secretion
          intestine smooth muscle contraction
          urinary bladder smooth muscle contraction
          negative regulation of cAMP biosynthetic process
          positive regulation of cAMP biosynthetic process
          positive regulation of fever generation
          response to estradiol
          response to progesterone
          response to drug
          positive regulation of vasoconstriction
          positive regulation of circadian sleep/wake cycle, non-REM sleep
          negative regulation of insulin secretion
          maternal process involved in parturition
          negative regulation of gastric acid secretion
          cellular response to lipopolysaccharide
          cellular response to interleukin-1
          negative regulation of platelet aggregation
          negative regulation of potassium ion transmembrane transport
          response to salt
          negative regulation of calcium ion transmembrane transport
          negative regulation of gastrin-induced gastric acid secretion
          positive regulation of smooth muscle contraction involved in micturition
          cellular response to forskolin
          positive regulation of inhibitory G-protein coupled receptor phosphorylation
          negative regulation of circadian sleep/wake cycle, wakefulness
          positive regulation of myofibroblast contraction
          positive regulation of colon smooth muscle contraction
          positive regulation of gastric mucosal blood circulation
          prostaglandin receptor internalization
          positive regulation of neutrophil extravasation
          negative regulation of forebrain neuron differentiation
          prostaglandin E receptor activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0