siRNA Details
Gene Symbol: CDCP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201546.3 | NP_963840.2 | 4 | 1334 |

Gene Symbol: CDCP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201546.3 | NP_963840.2 | 4 | 1334 |