siRNA Details
Gene Symbol: HHEX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002729.4 | NP_002720.1 | 2 | 542 | ||
GenBank mRNA | AK314891.1 | - | 2 | 511 | ||
BC014336.1 | - | 2 | 515 | 4046641 | ||
BC015110.1 | - | 2 | 515 | 4048691 | ||
BC050638.1 | - | 2 | 482 | 6174612 | ||
L16499.1 | - | 2 | 514 | |||
X67235.1 | - | 2 | 486 | |||
Z21533.1 | - | 1 | 457 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
