Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNB1IP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021178.4 | NP_067001.3 | 7 | 1414 | ||
NM_182849.2 | NP_878269.1 | 6 | 1214 | |||
NM_182852.3 | NP_878272.1 | 8 | 1595 | |||
GenBank mRNA | AK026233.1 | - | 7 | 1381 | ||
AK225253.1 | - | 7 | 1386 | |||
AK225451.1 | - | 7 | 1381 | |||
AK226037.1 | - | 7 | 1386 | |||
AL161994.1 | - | 7 | 1394 | |||
BC000369.2 | - | 6 | 1356 | 2819346 | ||
BC001218.1 | - | 6 | 1359 | 3528496 | ||
BC004435.1 | - | 6 | 1356 | 2819346 | ||
BX161423.1 | - | 7 | 1574 | |||
BX161424.1 | - | 7 | 1542 | |||
BX247978.1 | - | 7 | 1522 |