siRNA Details
Gene Symbol: PDXK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003681.4 | NP_003672.1 | 11 | 6396 | ||
XM_005261195.2 | XP_005261252.2 | 11 | 6495 | |||
XM_005261196.3 | XP_005261253.1 | 11 | 6240 | |||
XM_005261198.2 | XP_005261255.1 | 10 | 6245 | |||
XM_005261199.2 | XP_005261256.1 | 11 | 6242 | |||
XM_011529758.2 | XP_011528060.1 | 12 | 6785 | |||
XM_011529760.2 | XP_011528062.1 | 10 | 6097 | |||
XM_011529761.1 | XP_011528063.1 | 10 | 6109 | |||
XM_017028483.1 | XP_016883972.1 | 12 | 6542 | |||
XM_017028484.1 | XP_016883973.1 | 12 | 6872 | |||
GenBank mRNA | AK024977.1 | - | 1 | 828 | ||
AK027018.1 | - | 1 | 1616 | |||
AK225206.1 | - | 1 | 828 | |||
AL137632.1 | - | 1 | 2295 | |||
AL161960.1 | - | 1 | 3933 | |||
AL355712.1 | - | 1 | 260 | |||
AL355713.1 | - | 1 | 939 | |||
BC003651.2 | - | 1 | 298 | |||
EU794642.1 | - | 11 | 6396 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
