Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ELAVL4 SIRNA Assays ›
          Gene Symbol:
          ELAVL4
          Gene Name:
          ELAV like neuron-specific RNA binding protein 4
          Gene Aliases:
          HUD, PNEM
          Chromosome Location:
          Chr.1: 50048014 - 50203786 on Build GRCh38
          UniGene ID:
          Hs.213050
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 29400

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ELAVL4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001144774.2 NP_001138246.1 2, 3 528
          NM_001144775.2 NP_001138247.2 2, 3 378
          NM_001144776.2 NP_001138248.1 2, 3 340
          NM_001144777.2 NP_001138249.1 2, 3 418
          NM_001294348.1 NP_001281277.1 2, 3 598
          NM_001324208.1 NP_001311137.1 2, 3 418
          NM_001324209.1 NP_001311138.1 3, 4 615
          NM_001324212.1 NP_001311141.1 3, 4 615
          NM_001324213.1 NP_001311142.1 2, 3 598
          NM_001324214.1 NP_001311143.1 3, 4 685
          NM_001324215.1 NP_001311144.1 2, 3 598
          NM_001324216.1 NP_001311145.1 2, 3 528
          NM_001324217.1 NP_001311146.1 2, 3 598
          NM_021952.4 NP_068771.2 2, 3 528
          NR_136725.1 3, 4 685
          XM_006710411.3 XP_006710474.1 2, 3 510
          XM_011540889.2 XP_011539191.1 2, 3 393
          XM_011540890.2 XP_011539192.1 2, 3 392
          XM_011540893.2 XP_011539195.1 2, 3 340
          XM_011540894.2 XP_011539196.1 3, 4 682
          XM_011540895.2 XP_011539197.1 3, 4 586
          XM_017000538.1 XP_016856027.1 2, 3 512
          XM_017000539.1 XP_016856028.1 2, 3 1139
          XM_017000540.1 XP_016856029.1 2, 3 396
          XM_017000541.1 XP_016856030.1 2, 3 528
          XM_017000542.1 XP_016856031.1 2, 3 377
          GenBank mRNA AK297338.1 - 2, 3 418
          AK298817.1 - 2, 3 407
          AK298902.1 - 2, 3 598
          AK309653.1 - 2, 3 373
          AY033995.1 - 2, 3 340
          AY033996.1 - 2, 3 351
          AY033997.1 - 2, 3 515
          AY033998.1 - 2, 3 482
          BC036071.1 - 2, 3 486 5286347
          BM509527.1 - 2, 3 385
          DC351092.1 - 2, 3 418
          M62843.1 - 2, 3 352

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001077651, XM_006238593, XM_006238592, XM_006238591, XM_006238595, and others.
          This siRNA also targets the following transcripts in Mouse: NM_010488, NM_001163399, NM_001163397, NM_001038698, AK079088, and others.

          Panther Classification:

          Molecular Function -

          translation initiation factor

          Gene Ontology Categories:

          Function(s) Process(es)

          RNA processing
          mRNA processing
          nucleotide binding
          RNA binding
          mRNA 3'-UTR binding
          AU-rich element binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0