Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ELAVL4 SIRNA Assays ›
Gene Symbol:
ELAVL4
Gene Name:
ELAV like neuron-specific RNA binding protein 4
Gene Aliases:
HUD, PNEM
Chromosome Location:
Chr.1: 50048014 - 50203786 on Build GRCh38
UniGene ID:
Hs.213050
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
29490
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ELAVL4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001144774.2
NP_001138246.1
4
667
NM_001144775.2
NP_001138247.2
4
517
NM_001144776.2
NP_001138248.1
4
479
NM_001144777.2
NP_001138249.1
4
557
NM_001294348.1
NP_001281277.1
4
737
NM_001324208.1
NP_001311137.1
4
557
NM_001324209.1
NP_001311138.1
5
754
NM_001324212.1
NP_001311141.1
5
754
NM_001324213.1
NP_001311142.1
4
737
NM_001324214.1
NP_001311143.1
5
824
NM_001324215.1
NP_001311144.1
4
737
NM_021952.4
NP_068771.2
4
667
XM_006710411.3
XP_006710474.1
4
649
XM_011540889.2
XP_011539191.1
4
532
XM_011540890.2
XP_011539192.1
4
531
XM_011540893.2
XP_011539195.1
4
479
XM_011540894.2
XP_011539196.1
5
821
XM_011540895.2
XP_011539197.1
5
725
XM_017000538.1
XP_016856027.1
4
651
XM_017000539.1
XP_016856028.1
4
1278
GenBank mRNA
AK297338.1
-
4
557
AK298817.1
-
4
546
AK298902.1
-
4
737
AK309653.1
-
4
512
AY033995.1
-
4
479
AY033996.1
-
4
490
AY033997.1
-
4
654
AY033998.1
-
4
621
BC036071.1
-
4
625
5286347
DC351092.1
-
4
557
M62843.1
-
4
491
Back To Top
More Information
Panther Classification:
Molecular Function -
translation initiation factor
Gene Ontology Categories:
Function(s)
Process(es)
RNA processing
mRNA processing
nucleotide binding
RNA binding
mRNA 3'-UTR binding
AU-rich element binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0