Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C12orf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021640.3 | NP_067653.3 | 6 | 879 | ||
GenBank mRNA | AF111805.1 | - | 4 | 471 | ||
AF289485.1 | - | 6 | 847 | |||
BC013956.2 | - | 5 | 666 | |||
BC028904.1 | - | 5 | 783 | 4454943 | ||
BC051871.1 | - | 6 | 838 | 5170816 | ||
BM551107.1 | - | 2 | 185 |