Search
Search

GALNT11| Gene Symbol: GALNT11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022087.3 | NP_071370.2 | 2 | 525 | ||
| XM_006716082.2 | XP_006716145.1 | 2 | 436 | |||
| XM_006716083.2 | XP_006716146.1 | 2 | 404 | |||
| XM_006716084.2 | XP_006716147.1 | 2 | 368 | |||
| XM_017012499.1 | XP_016867988.1 | 2 | 506 | |||
| XM_017012500.1 | XP_016867989.1 | 2 | 506 | |||
| GenBank mRNA | AK025287.1 | - | 2 | 359 | ||
| AK296097.1 | - | 2 | 382 | |||
| BC059377.1 | - | 3 | 631 | |||
| DA308896.1 | - | 2 | 448 | |||
| DA679635.1 | - | 2 | 395 | |||
| Y12434.3 | - | 1 | 276 | |||