Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POPDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308333.1 | NP_001295262.1 | 1 | 493 | ||
NM_022135.3 | NP_071418.2 | 1 | 493 | |||
XM_005247698.3 | XP_005247755.1 | 1 | 493 | |||
GenBank mRNA | AF204173.1 | - | 1 | 476 | ||
AK226022.1 | - | 2 | 558 | |||
AK314819.1 | - | 1 | 494 | |||
BC044929.1 | - | 1 | 493 | 5191230 |