Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CDH23 SIRNA Assays ›
          Gene Symbol:
          CDH23
          Gene Name:
          cadherin-related 23
          Gene Aliases:
          CDHR23, USH1D
          Chromosome Location:
          Chr.10: 71396934 - 71815947 on Build GRCh38
          UniGene ID:
          Hs.656032
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 29754

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CDH23
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001171930.1 NP_001165401.1 9, 10 1230
          NM_001171931.1 NP_001165402.1 9, 10 1230
          NM_001171932.1 NP_001165403.1 9, 10 1230
          NM_022124.5 NP_071407.4 9, 10 1230
          NM_052836.3 NP_443068.1 9, 10 1230
          XM_006717940.3 XP_006718003.1 9, 10 1224
          XM_006717942.3 XP_006718005.1 9, 10 1224
          XM_011540039.2 XP_011538341.1 9, 10 1224
          XM_011540042.2 XP_011538344.1 9, 10 1224
          XM_011540043.2 XP_011538345.1 9, 10 1224
          XM_011540044.2 XP_011538346.1 9, 10 865
          XM_011540045.2 XP_011538347.1 9, 10 1224
          XM_011540046.2 XP_011538348.1 4, 5 659
          XM_011540048.2 XP_011538350.1 9, 10 1224
          XM_011540049.2 XP_011538351.1 9, 10 1224
          XM_011540051.2 XP_011538353.1 9, 10 1224
          XM_017016499.1 XP_016871988.1 9, 10 1230
          XM_017016500.1 XP_016871989.1 9, 10 1224
          GenBank mRNA AF312024.1 - 8, 9 840
          AY010111.2 - 8, 9 840
          AY358617.1 - 9, 10 880
          BC011570.1 - 9, 10 1235
          BC032581.1 - 9, 10 1236
          BC065284.1 - 9, 10 1207
          BC108254.1 - 7, 8 998
          BC136976.1 - 9, 10 1100
          BC136977.1 - 9, 10 1099
          BC139903.1 - 9, 10 1222

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_001252635, NM_023370, EU681829, AJ888000, AJ888002, and others.

          Panther Classification:

          Molecular Function -

          cadherin

          Gene Ontology Categories:

          Function(s) Process(es)

          calcium ion transport
          homophilic cell adhesion via plasma membrane adhesion molecules
          visual perception
          sensory perception of sound
          locomotory behavior
          calcium-dependent cell-cell adhesion via plasma membrane cell adhesion molecules
          photoreceptor cell maintenance
          response to stimulus
          sensory perception of light stimulus
          equilibrioception
          regulation of cytosolic calcium ion concentration
          inner ear receptor stereocilium organization
          calcium ion binding
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0