Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPB41L4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022140.3 | NP_071423.3 | 9 | 1041 | ||
XM_011543530.1 | XP_011541832.1 | 9 | 1218 | |||
XM_011543531.2 | XP_011541833.1 | 9 | 1218 | |||
XM_011543532.1 | XP_011541834.1 | 9 | 1218 | |||
XM_011543533.1 | XP_011541835.1 | 9 | 1218 | |||
XM_017009689.1 | XP_016865178.1 | 9 | 1218 | |||
GenBank mRNA | AB030240.1 | - | 9 | 770 | ||
AK123285.1 | - | 5 | 1768 | |||
BC114632.1 | - | 9 | 867 | |||
BC114942.1 | - | 9 | 862 |