Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031714.3 | NP_001026884.3 | 20 | 3055 | ||
NM_022489.3 | NP_071934.3 | 20 | 3055 | |||
XM_005268004.4 | XP_005268061.1 | 20 | 3142 | |||
XM_005268005.4 | XP_005268062.1 | 20 | 3113 | |||
XM_017021595.1 | XP_016877084.1 | 20 | 3113 | |||
GenBank mRNA | AF318379.1 | - | 2 | 139 | ||
AK025709.1 | - | 11 | 1018 | |||
AK225325.1 | - | 11 | 1018 | |||
BC008756.2 | - | 10 | 963 | |||
BC064828.1 | - | 4 | 373 | |||
BU538960.1 | - | 6 | 531 | |||
CD631414.1 | - | 2 | 54 | |||
DQ395338.1 | - | 13 | 1316 | |||
DQ395339.1 | - | 13 | 1316 | |||
DQ395340.1 | - | 13 | 1316 |